Learn More
Grainger GRADUATED CYLINDER, PP, 2000ML CAP
Catalog No. NC2661599
Encompass_Preferred
The 2000ml polypropylene graduated cylinder by Globe Scientific is made from clear polypropylene plastic. It has a wide base for stability and features large, easy-to-read graduations in both metric and US customary units. This cylinder has a pouring lip and a hexagonal base to prevent rolling. It can withstand temperatures from -20°C to 80°C. The 2000ml capacity cylinder has an accuracy of ±2%. This durable graduated cylinder is autoclavable and ideal for general purpose liquid measuring in laboratories.
Catalog No. NC2661599 Supplier Grainger Supplier No. 8ADU8
The 2000ml polypropylene graduated cylinder by Globe Scientific is made from clear polypropylene plastic. It has a wide base for stability and features large, easy-to-read graduations in both metric and US customary units. This cylinder has a pouring lip and a hexagonal base to prevent rolling. It can withstand temperatures from -20°C to 80°C. The 2000ml capacity cylinder has an accuracy of ±2%. This durable graduated cylinder is autoclavable and ideal for general purpose liquid measuring in laboratories.