Learn More
Grainger LAB SAFETY SUPPLY, Graduated Cylinder, Polypropylene
Catalog No. NC2661606
Encompass_Preferred
The Lab Safety Supply Graduated Cylinder is made of clear polypropylene plastic with easy-to-read black graduations. It has a capacity of 100 mL with graduations of 1 mL. The cylinder has a hexagonal base to prevent rolling and has a pour spout for easy, accurate dispensing. It can withstand temperatures from -20°C to 80°C. The dimensions are 3-1/2" diameter x 5-7/8" height. This durable graduated cylinder is autoclavable and resistant to acids, bases, and most solvents. It meets ASTM E694 Class A specifications.
Catalog No. NC2661606 Supplier Grainger Supplier No. 9WH45
The Lab Safety Supply Graduated Cylinder is made of clear polypropylene plastic with easy-to-read black graduations. It has a capacity of 100 mL with graduations of 1 mL. The cylinder has a hexagonal base to prevent rolling and has a pour spout for easy, accurate dispensing. It can withstand temperatures from -20°C to 80°C. The dimensions are 3-1/2" diameter x 5-7/8" height. This durable graduated cylinder is autoclavable and resistant to acids, bases, and most solvents. It meets ASTM E694 Class A specifications.