Learn More
MSC Dynalon Labware Graduated Cylinder, Polypropylene, 4 L, 1 ea
Catalog No. 501909394
Encompass
The graduated cylinder, made from polypropylene, offers accurate liquid measurement and is designed for laboratory use. It features a removable base for ease of cleaning and storage, and it can be autoclaved for sterilization.
  • Autoclavable for sterilization
  • Removable base for cleaning and storage
  • Transparent for clear viewing of liquid levels
  • Complies with US FDA 21 CFR 177.1520 and ISO 6706 standards
Catalog No. 50-190-9394 Supplier MSC Supplier No. 35674217
The graduated cylinder, made from polypropylene, offers accurate liquid measurement and is designed for laboratory use. It features a removable base for ease of cleaning and storage, and it can be autoclaved for sterilization.
  • Autoclavable for sterilization
  • Removable base for cleaning and storage
  • Transparent for clear viewing of liquid levels
  • Complies with US FDA 21 CFR 177.1520 and ISO 6706 standards