Learn More
Uline GRAD MIXING PAIL 5 GA -MIN 5EA
Catalog No. NC3662383
Encompass
Graduated Mixing Pail 5 GAL - MINIMUM 5 EACHGraduation markings in gallons and liters.Made of translucent, high-density polyethylene.Withstands temperatures up to 180°F.FDA compliant.
Catalog No. NC3662383 Supplier Uline Supplier No. S24266
Graduated Mixing Pail 5 GAL - MINIMUM 5 EACHGraduation markings in gallons and liters.Made of translucent, high-density polyethylene.Withstands temperatures up to 180°F.FDA compliant.