Learn More
MSC Value Collection Square Steel Gage Block, 0.147 in., Grade 0, 1 ea.
Catalog No. 5000547834
Encompass
This high-precision square steel gage block is designed for accurate dimensional measurements and calibration tasks. Engineered for stability and reliability, it meets Grade 0 accuracy standards for demanding metrology applications.
  • Constructed from high-quality steel for durability
  • Square design facilitates secure stacking and handling
  • Certified with NIST traceability to ensure measurement accuracy
  • Grade 0 accuracy for high-precision requirements
Catalog No. 50-005-47834 Supplier MSC Supplier No. 86273356
This high-precision square steel gage block is designed for accurate dimensional measurements and calibration tasks. Engineered for stability and reliability, it meets Grade 0 accuracy standards for demanding metrology applications.
  • Constructed from high-quality steel for durability
  • Square design facilitates secure stacking and handling
  • Certified with NIST traceability to ensure measurement accuracy
  • Grade 0 accuracy for high-precision requirements