Learn More
MSC Value Collection Gage Block, Square Steel, Grade 0, 0.17 in., 1 ea.
Catalog No. 5000516006
Encompass
This 0.17 in. square gage block is engineered from high-quality steel to provide reliable measurement and calibration standards. Designed for precision applications, it meets grade 0 accuracy requirements for exacting industrial workflows.
  • Accuracy grade 0
  • Crafted from durable steel
  • Square configuration
  • Made in the USA
Catalog No. 50-005-16006 Supplier MSC Supplier No. 86273703
This 0.17 in. square gage block is engineered from high-quality steel to provide reliable measurement and calibration standards. Designed for precision applications, it meets grade 0 accuracy requirements for exacting industrial workflows.
  • Accuracy grade 0
  • Crafted from durable steel
  • Square configuration
  • Made in the USA