Learn More
MSC Value Collection Square Steel Gage Block, 0.115 in., Grade 0, 1 ea.
Catalog No. 5000515675
Encompass
This square gage block is designed for precise measurement and calibration applications in metalworking and machining environments. Constructed from high-quality steel, this block provides reliable dimensional accuracy for layout and machine setup tasks.
  • Accuracy grade 0 for high-precision measurement
  • Square shape for versatile positioning
  • Constructed from durable steel
  • Includes NIST traceability certification
Catalog No. 50-005-15675 Supplier MSC Supplier No. 86271806
This square gage block is designed for precise measurement and calibration applications in metalworking and machining environments. Constructed from high-quality steel, this block provides reliable dimensional accuracy for layout and machine setup tasks.
  • Accuracy grade 0 for high-precision measurement
  • Square shape for versatile positioning
  • Constructed from durable steel
  • Includes NIST traceability certification