Learn More
MSC Value Collection Square Steel Gage Block, 0.1003 in., Grade 0, 1 ea.
Catalog No. 5000515288
Encompass
This square steel gage block is designed for precise measurement and calibration applications. Constructed from durable steel, this gage block ensures reliability in demanding environments.
  • Accuracy grade 0
  • Size: 0.1003 in.
  • Includes NIST traceability certification
  • Origin: USA
Catalog No. 50-005-15288 Supplier MSC Supplier No. 86271038
This square steel gage block is designed for precise measurement and calibration applications. Constructed from durable steel, this gage block ensures reliability in demanding environments.
  • Accuracy grade 0
  • Size: 0.1003 in.
  • Includes NIST traceability certification
  • Origin: USA