Learn More
Chemglass Life Sciences Cylinder, Graduated, 100mL, Single Scale, To Contain
SDP
Catalog No. NC1448466
Encompass_Preferred
Calibrated ¿To Contain¿, with white enamel graduations. The 100mL capacity is in accordance with ASTM E-133 and E-1272 and is used in ASTM tests D-86, D-216 and D-447.
Catalog No. NC1448466 Supplier Chemglass Life Sciences Supplier No. CG8238100
Calibrated ¿To Contain¿, with white enamel graduations. The 100mL capacity is in accordance with ASTM E-133 and E-1272 and is used in ASTM tests D-86, D-216 and D-447.