Learn More
Chemglass Life Sciences Cylinder, Economy, 10mL, Graduated, Double Scale
SDP
Catalog No. NC0175349
Encompass_Preferred
Calibrated "To Contain". A less costly cylinder, designed for those institutions performing many general laboratory procedures. No bumper guards are supplied.
This item is not returnable. View return policy
Catalog No. NC0175349 Supplier Chemglass Life Sciences Supplier No. CG824610
Calibrated "To Contain". A less costly cylinder, designed for those institutions performing many general laboratory procedures. No bumper guards are supplied.