Learn More
Chemglass Life Sciences Cylinder, Graduated, 10mL, Single Scale, To Contain
SDP
Catalog No. NC0727568
Encompass_Preferred
Cylinder, Graduated, 10mL, Single Scale, To Contain with white enamel graduations. Has a funnel top.
Catalog No. NC0727568 Supplier Chemglass Life Sciences Supplier No. CG823810
Cylinder, Graduated, 10mL, Single Scale, To Contain with white enamel graduations. Has a funnel top.