Learn More
Cole-Parmer Thermo Scientific Nalgene 3665-0100 graduated polymethylpentene cylinder, 100 mL
Catalog No. NC0535769 Shop All Cole-Parmer Products
Encompass_Preferred
Transparent polymethylpentene (PMP) cylinders with large pour spout offer good chemical and heat resistance. Easy-to-read, raised graduations allow precise measuring. Autoclavable.
Catalog No. NC0535769 Supplier Cole-Parmer Supplier No. 0613445
Transparent polymethylpentene (PMP) cylinders with large pour spout offer good chemical and heat resistance. Easy-to-read, raised graduations allow precise measuring. Autoclavable.