Learn More
Chemglass Life Sciences Chemglass Lifetime Red Pyrex graduated cylinder, 50 mL, single metric scale
SDP
Catalog No. NC2583348
Encompass_Preferred
Chemglass Lifetime Red PYREX graduated cylinder provides reliable 50 mL liquid measurement with a single metric scale and reinforced bead to help reduce breakage. It is suitable for routine laboratory measurement and transfer.
  • Capacity: 50 mL
  • Material: borosilicate glass
  • Color: red stripe
  • Scale: single metric scale
  • Feature: reinforcement bead for added durability
  • Calibration: to contain
  • Packaging: single each
Catalog No. NC2583348 Supplier Chemglass Life Sciences Supplier No. CG825450
Chemglass Lifetime Red PYREX graduated cylinder provides reliable 50 mL liquid measurement with a single metric scale and reinforced bead to help reduce breakage. It is suitable for routine laboratory measurement and transfer.
  • Capacity: 50 mL
  • Material: borosilicate glass
  • Color: red stripe
  • Scale: single metric scale
  • Feature: reinforcement bead for added durability
  • Calibration: to contain
  • Packaging: single each