Learn More
Chemglass Life Sciences Chemglass Graduated Cylinder, Class B, 500 mL, Borosilicate Glass, With Bumper
SDP
Catalog No. NC2590714
Encompass_Preferred
The Chemglass Class B 500 mL graduated cylinder is a borosilicate glass measuring cylinder calibrated "to deliver" and supplied with a protective bumper to reduce breakage and improve handling.
  • 500 mL capacity for routine laboratory measurements.
  • Borosilicate glass (Type 3.3) for chemical resistance and thermal stability.
  • Class B accuracy compliant with ASTM Specification E1272 for reliable measurements.
  • White single metric scale with clear ring-molded graduations for easy reading.
  • Protective bumper reduces breakage and improves grip during handling.
  • Calibrated "to deliver" for accurate dispensing.
Catalog No. NC2590714 Supplier Chemglass Life Sciences Supplier No. CG8242K500
The Chemglass Class B 500 mL graduated cylinder is a borosilicate glass measuring cylinder calibrated "to deliver" and supplied with a protective bumper to reduce breakage and improve handling.
  • 500 mL capacity for routine laboratory measurements.
  • Borosilicate glass (Type 3.3) for chemical resistance and thermal stability.
  • Class B accuracy compliant with ASTM Specification E1272 for reliable measurements.
  • White single metric scale with clear ring-molded graduations for easy reading.
  • Protective bumper reduces breakage and improves grip during handling.
  • Calibrated "to deliver" for accurate dispensing.