Learn More
Hach Company Graduated Cylinder Low 100mL
Supplier:  Hach Company 2928402
Encompass
Cylinder-Graduated-Double Metric-Low Form-100 mLThese graduated cylinders feature two sets of graduations for reading either up or down and two pouring Iips Calibrated in to deliverin
Catalog No. 50-002-07696