Learn More
Hach Company Graduated Cylinder PMP 500 mL
Supplier:  Hach Company 217249
Encompass
This cylinder features transparent nontoxic noncontaminating and biologically inert material with excellent chemical resistance Meets ASTM E1272 Class B accuracy requirements ISO Standard 6706 and suitable for food and beverage use (PMP resin meets requirements of CFR 21 Part 177 1520) Calibrated To Contain/To Deliver at 20 C (68 F) Shatterproof and thermally resistant to 135 C (275 F) Absence of meniscus for easy-to-read levels on front-to-back raised graduations
Catalog No. 50-002-57141