Learn More
Hach Company Graduated Cylinder PP 50mL
Supplier:  Hach Company 108141
Encompass
Graduated Cylinder polypropylene 50 mL -0 6 mL with 1 0 mL divisionsPolypropylene construction of both cylinder and base All sizes have generous pour spout large molded-in and easy-to-read graduations Can be chemically sterilized without affecting accuracy Autoclavable but accuracy will be adversely affected Cylinders meet accuracy requirements of ASTM Class B E1272 ISO Standard 6706 and CFR 21 Part 177 1520
Catalog No. 50-002-57102