Learn More
Alkali Scientific Graduated Cylinders, Polypropylene (PP), Molded Graduations, 500mL, 8/Case
Supplier: Alkali Scientific MC500
These economy graduated cylinders are manufactured from durable polypropylene (PP) and are designed for routine liquid measurement in laboratory, educational, and research environments. Permanently molded graduations provide clear volume markings that will not fade, chip, or wash away over time. The light and impact resistant polypropylene construction offers good chemical resistance and makes these cylinders a practical alternative to glass for everyday laboratory use. The economy design provides reliable performance while maintaining cost efficiency. Available in volumes from 10 mL to 500 mL, these cylinders are suitable for sample preparation, reagent measurement, and general laboratory liquid handling procedures.