Learn More
Hach Company Graduated mixing cyln 100mL
Supplier:  Hach Company 103826
Encompass
Graduated Cylinder for mixing Glass 100 mL -0 6 mL with 1 0 mL divisions and glass stopper 16This glass graduated cylinder is calibrated to contain with a molded in base Each includes glass penny head stopper Graduations are in durable white enamel bake
Catalog No. 50-002-57128