Learn More
Electron Microscopy Sciences Graduated Pitcher, 3000 ml, 1 each
SDP
Catalog No. 5026331
Encompass_Preferred
Gracefully proportioned, molded pitchers with large comfortable handles, metric graduations and well designed pouring spouts. They resist heat to 175 degree C (374 degree F), are shatter-proof and have excellent chemical resistance. Ideal for preparation of photographic chemicals.
Catalog No. 50-263-31 Supplier Electron Microscopy Sciences Supplier No. 74372
Gracefully proportioned, molded pitchers with large comfortable handles, metric graduations and well designed pouring spouts. They resist heat to 175 degree C (374 degree F), are shatter-proof and have excellent chemical resistance. Ideal for preparation of photographic chemicals.