Learn More
Marketlab, Inc. Graduated Pitcher
Description
- Raised numerals and non-wetting surface for easy reading
- Flared rims and large spouts for easy pouring
- Resistant to most commonly used lab reagents
- Autoclavable at 121°C (250°F)
- Graduation interval: 250 mL
Specifications
Specifications
| Capacity (Metric) | 5000 mL |
| Color | Clear |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Closure Included | No |
| Graduated | Yes |
| Mouth | Wide |
| Autoclavable | Autoclavable |
| Capacity (English) | 169.07 oz. |
| Shape | Tapered |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.