Learn More
Thomas Scientific Graduated, PC Collection Bottle
SDP
Catalog No. 501461416
Encompass_Preferred
The Accuris ASPIRE Laboratory Aspirator incorporates a quiet, maintenance-free pump inside a sturdy, yet compact base. The vacuum is fully adjustable, with a pressure gauge that displays the vacuum level, allowing full control of aspirating speed. The ASPIRE is an ideal choice for a wide range of laboratory aspirating applications, including removal of media from tissue culture vessels or plates, and supernatants. An ergonomic, hand held aspirating device is included, and holds standard 200 to 300ul disposable pipette tips. Comes complete with: Base with internal vacuum pump 2 Liter, Polycarbonate Collection Bottle (autoclavable and break resistant) Antibacterial / Hydrophobic Filter Silicone Tubing Handheld Vacuum Pipette 2 Pipette cone adapters The collection bottle incorporates a reliable Overflow Protection Valve that closes off the tube to the vacuum in the event of a full bottle. In addition, the replaceable in-line filter protects the vacuum pump from contaminants and any moistu
Catalog No. 50-146-1416 Supplier Thomas Scientific Supplier No. 1191L48
The Accuris ASPIRE Laboratory Aspirator incorporates a quiet, maintenance-free pump inside a sturdy, yet compact base. The vacuum is fully adjustable, with a pressure gauge that displays the vacuum level, allowing full control of aspirating speed. The ASPIRE is an ideal choice for a wide range of laboratory aspirating applications, including removal of media from tissue culture vessels or plates, and supernatants. An ergonomic, hand held aspirating device is included, and holds standard 200 to 300ul disposable pipette tips. Comes complete with: Base with internal vacuum pump 2 Liter, Polycarbonate Collection Bottle (autoclavable and break resistant) Antibacterial / Hydrophobic Filter Silicone Tubing Handheld Vacuum Pipette 2 Pipette cone adapters The collection bottle incorporates a reliable Overflow Protection Valve that closes off the tube to the vacuum in the event of a full bottle. In addition, the replaceable in-line filter protects the vacuum pump from contaminants and any moistu