Learn More
Synthware™ Graduated Single Tube Receiver
GREENER_CHOICESDP
Catalog No. 31501035 Shop All Kemtech Products
Change view
Click to view available options
Capacity (Metric):
12 mL
15 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
31-501-035 15 mL
31-501-034 12 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-501-035 Supplier Synthware™ Supplier No. T230015
Add to Cart
Add to Cart
Centrifuge tube shaped receiver with an upper standard taper joint.
Specifications
For Use With (Application) Organic Synthesis
Type Distillation Receiver
Material Borosilicate Glass
Capacity (Metric) 15 mL
Quantity 1 Ea.
Standard Taper Joint 14/20