Learn More
Andwin Scientific GRADUATED TO 1.0 ML STERILIZED
SDP
Catalog No. 50819423
Encompass_Preferred
The 5 mL sterile transfer pipet is made of polyethyelene and therefore will not shatter or break like those that are made of glass. This product is an ideal alternative to pasteur pipets and rubber bulbs. The transfer pipet has an overall length of 15 centimeters and graduations at 0.25, 0.5, 0.75, and 1.0 mL. Sold in a pack of 400 individually wrapped pipets.
Catalog No. 50-819-423 Supplier Andwin Scientific Supplier No. 10222510
The 5 mL sterile transfer pipet is made of polyethyelene and therefore will not shatter or break like those that are made of glass. This product is an ideal alternative to pasteur pipets and rubber bulbs. The transfer pipet has an overall length of 15 centimeters and graduations at 0.25, 0.5, 0.75, and 1.0 mL. Sold in a pack of 400 individually wrapped pipets.