Learn More
Andwin Scientific GRADUATED TO 1.0 ML STERILIZED
SDP
Supplier:  Andwin Scientific 10222520
Encompass_Preferred
The 5 mL sterile transfer pipet is made of polyethyelene and therefore will not shatter or break like those that are made of glass. This product is an ideal alternative to pasteur pipets and rubber bulbs. The transfer pipet has an overall length of 15 centimeters and graduations at 0.25, 0.5, 0.75, and 1.0 mL. Sold in a pack of 400.
Catalog No. 50-819-424