Learn More
Andwin Scientific GRADUATED TO 3.0 ML TRANSFER P
SDP
Catalog No. 50819415
Encompass_Preferred
The 7 mL transfer pipet is made of polyethyelene and therefore will not shatter or break like those that are made of glass. This product is an ideal alternative to pasteur pipets and rubber bulbs. The transfer pipet has an overall length of 15.5 centimeters and graduations at 0.5, 1.0, 2.0, and 3.0 mL. Sold in a pack of 500.
Catalog No. 50-819-415 Supplier Andwin Scientific Supplier No. 10202500
The 7 mL transfer pipet is made of polyethyelene and therefore will not shatter or break like those that are made of glass. This product is an ideal alternative to pasteur pipets and rubber bulbs. The transfer pipet has an overall length of 15.5 centimeters and graduations at 0.5, 1.0, 2.0, and 3.0 mL. Sold in a pack of 500.