Learn More
Research Products International Corp Graduated Transfer Pipette, Sterile, Individually Wrapped, 500 per Package
SDP
Catalog No. 50998530
Encompass
Plastic transfer pipettes are molded into one-piece units from low density polyethylene. They eliminate cross-contamination encountered with rubber bulbs and reduce exposure when working with hazardous or infectious samples. Ideal for all laboratory procedures that require fast and safe transferring of liquids. Pipettes can also be used as freeze vials or as storage and transport containers by merely heat sealing the tips closed after filling.
Graduated: 1ml x 0.25ml increments. Stem tapers to a fine tip. Ideal for specialized laboratory procedures, storage or transporting of samples.
Catalog No. 50-998-530 Supplier Research Products International Corp Supplier No. 1475011S
Plastic transfer pipettes are molded into one-piece units from low density polyethylene. They eliminate cross-contamination encountered with rubber bulbs and reduce exposure when working with hazardous or infectious samples. Ideal for all laboratory procedures that require fast and safe transferring of liquids. Pipettes can also be used as freeze vials or as storage and transport containers by merely heat sealing the tips closed after filling.
Graduated: 1ml x 0.25ml increments. Stem tapers to a fine tip. Ideal for specialized laboratory procedures, storage or transporting of samples.