Learn More
Sigma Aldrich Fine Chemicals Biosciences Graduated ValumetricTM bot
Catalog No. 502905659
Encompass
Ideal for use as sample bottles. They are also useful for dry powders soil samples and specimen storage. All bottles are made of clear Type III soda lime glass.
Catalog No. 50-290-5659 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5510071PAK
Ideal for use as sample bottles. They are also useful for dry powders soil samples and specimen storage. All bottles are made of clear Type III soda lime glass.