Learn More
Electron Microscopy Sciences Graefe Dissecting Knife, 27 mm x 1.7 mm, 4¾” OAL
SDP
Catalog No. 5034610
Encompass_Preferred
4 3/4" long, double edge blade
Blade: 27 mm x 1.7 mm
Catalog No. 50-346-10 Supplier Electron Microscopy Sciences Supplier No. 7793402
4 3/4" long, double edge blade
Blade: 27 mm x 1.7 mm