Learn More
Roboz Surgical GRAEFE FCPS 2.75 IN SERR CRV
SDP
Catalog No. NC9783984
Encompass
Graefe Forceps; Serrated, Curved; 0.6mm Tip Width; 2.75 in. Length
Catalog No. NC9783984 Supplier Roboz Surgical Supplier No. RS5111
Graefe Forceps; Serrated, Curved; 0.6mm Tip Width; 2.75 in. Length