Learn More
World Precision Instrument Graefe Fixation Forceps, 11.5cm, 5x5 Teeth, 4mm Tip
SDP
Catalog No. 502521363
Encompass
  • 11.5cm (4.5") Long
  • Straight
  • 4mm Tips
  • 5x5 Teeth
  • Stainless steel
Catalog No. 50-252-1363 Supplier World Precision Instrument Supplier No. 501988
  • 11.5cm (4.5") Long
  • Straight
  • 4mm Tips
  • 5x5 Teeth
  • Stainless steel