Learn More
Fine Science Tools Graefe Forceps, 10cm
SDP
Supplier:  Fine Science Tools 1165210
Encompass
Tip Shape: Curved/ Tips: Serrated/ Tip Dimensions: 1 x 0.9mm/ Alloy / Material: Titanium/ Length: 10cm
Catalog No. NC0175041