Learn More
Fine Science Tools Graefe Forceps, 10cm
SDP
Catalog No. NC0431920
Encompass
Tip Shape: Curved/ Tips: Serrated/ Tip Dimensions: 1 x 0.9mm/ Alloy / Material: Titanium/ Length: 10cm
Catalog No. NC0431920 Supplier Fine Science Tools Supplier No. 1165110
Tip Shape: Curved/ Tips: Serrated/ Tip Dimensions: 1 x 0.9mm/ Alloy / Material: Titanium/ Length: 10cm