Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Detco Graffiti Remover, 32 oz, Liquid, Biodegradable, 1 ea

Catalog No. 502868579
Encompass

The Detco Citrus Tagr is a biodegradable, citrus-based liquid designed to effectively remove tar, asphalt, and graffiti. It is suitable for use on surfaces such as cinder blocks, brick, and concrete.

  • Pleasant orange fragrance
  • Biodegradable formula
  • Non-hazardous composition
  • Sprayer bottle with 32 oz capacity
  • VOC content of 533 g/L
  • Flash point of 140 °F

Catalog No. 50-286-8579 Supplier MSC Supplier No. 33685462
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Detco Citrus Tagr is a biodegradable, citrus-based liquid designed to effectively remove tar, asphalt, and graffiti. It is suitable for use on surfaces such as cinder blocks, brick, and concrete.

  • Pleasant orange fragrance
  • Biodegradable formula
  • Non-hazardous composition
  • Sprayer bottle with 32 oz capacity
  • VOC content of 533 g/L
  • Flash point of 140 °F

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.