Learn More
MSC Bare Ground Solutions Graffiti Remover and Cleaner, 28 oz, Spray Bottle
Catalog No. 502838612
Encompass
This 100% biodegradable, all-natural graffiti remover effectively cleans grease, spray paint, tar, markers, and pencil from hard surfaces, including mold and mildew from tiles and grout. Non-flammable, with no harsh fumes or acids, it's VOC compliant and non-toxic. Its vertical cling prevents dripping and restaining on surfaces.
  • Removes grease, spray paint, tar, markers, and pencil from hard surfaces.
  • 100% biodegradable and all-natural.
  • VOC compliant and non-toxic.
  • Non-flammable, no harsh fumes or acids.
  • Vertical cling and dwell prevent dripping, running, and restaining.
  • Cleans mold and mildew from tiles and grout.
  • Eliminates Calcium Chloride buildup.
  • Safe for marine, auto, and RV use.
  • Easy-to-use trigger sprayer.
  • Safe on plastics, upholstery, carpet, and stainless steel.
Catalog No. 50-283-8612 Supplier MSC Supplier No. 50696616
This 100% biodegradable, all-natural graffiti remover effectively cleans grease, spray paint, tar, markers, and pencil from hard surfaces, including mold and mildew from tiles and grout. Non-flammable, with no harsh fumes or acids, it's VOC compliant and non-toxic. Its vertical cling prevents dripping and restaining on surfaces.
  • Removes grease, spray paint, tar, markers, and pencil from hard surfaces.
  • 100% biodegradable and all-natural.
  • VOC compliant and non-toxic.
  • Non-flammable, no harsh fumes or acids.
  • Vertical cling and dwell prevent dripping, running, and restaining.
  • Cleans mold and mildew from tiles and grout.
  • Eliminates Calcium Chloride buildup.
  • Safe for marine, auto, and RV use.
  • Easy-to-use trigger sprayer.
  • Safe on plastics, upholstery, carpet, and stainless steel.