Learn More
Med Vet International Graft Passer with Small Eye, J0236SE
Catalog No. 5000585308
Encompass_Preferred
Instrument with handleand thin 3cm radius curved tip with small eye. Thesmall eye will hold the large nylon suture used incruciate repair. The curved end is used to accesstight areas (i.e., between the fabella and femuralcondyle). Stainless steel. 6"L.
Catalog No. 50-005-85308 Supplier Med Vet International Supplier No. JOR236SE
Instrument with handleand thin 3cm radius curved tip with small eye. Thesmall eye will hold the large nylon suture used incruciate repair. The curved end is used to accesstight areas (i.e., between the fabella and femuralcondyle). Stainless steel. 6"L.