Learn More
Graham-Field™ Grafco™ Berman Type Oral Airways

Description
- Reinforced bite block to prevent airway collapse
- Single patient use recommended
- Choice of multi-size box or infant size pack
- Gas/cold sterilizable
- Latex free
Specifications
Specifications
| Product Type | Oral Airway |
| Color | White |
| Material | Polypropylene |
| Includes | Reinforced bite block |
| For Use With (Application) | For single patient use |
| Latex Free | Yes |
| Size | Infant, 50 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.