Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Graham-Field™ Wallmax Aneroid Sphygmomanometers
SDP

Catalog No. 19065890
Change view
Click to view available options
Size:
Adult
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-065890 Adult
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-065890 Supplier Graham-Field™ Supplier No. 222

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Size Adult
For Use With (Application) To measure arterial blood pressure
Includes Gauge, swivel bracket and mounting screws
Product Type Aneroid Sphygmomanometer
Color Blue
Width (Metric) Cuff 52.07cm
Width (English) Cuff 20.5 in.
Height (English) Cuff 5.5 in.
Height (Metric) Cuff 13.97cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.