Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Premium Grain Cowhide Leather Drivers Gloves - Straight Thumb

Catalog No. 17200393 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-200-393 X-Large
17-200-390 Small
17-200-391 Medium
17-200-392 Large
17-200-394 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-200-393 Supplier Wells Lamont™ Supplier No. Y0122XL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Premium Grain Cowhide Leather Drivers Gloves with Straight Thumb are constructed from premium grade leather to provide comfort and protection while on the job.

  • Double shirred elastic wrist
  • Bound hem
  • Straight thumb
  • Gunn cut

Specifications

Cuff Material Elastic
Color Natural
Grade Premium
Hand Specific
Latex Free Yes
Material Grain Cowhide Leather
Product Type Driver Gloves
Size X-Large
Style Gunn
Glove Cuff Color Blue
Glove Cuff Style Open
Hem Color Blue
Series Y0122
Thumb Style Straight
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.