Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grain Ethanol, 200 Proof, Ricca Chemical Company
SDP

Catalog No. RSOE00135E4 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
5 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RSOE00135E4 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RSOE00135E4 Supplier Ricca Chemical Company Supplier No. RSOE00135E4
Add to Cart
Add to Cart

Grain Ethanol is an essential solvent for pharmaceuticals, personal care products, laboratories, and industrial processes.

  • Ethanol, Grain
  • 200 Proof
  • 5 Gallon Metal Drum
  • High Purity Solvent
  • Essential across all industries

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
IUPAC Name ethanol
SMILES CCO

Specifications

Quantity 5 gal.
Packaging Metal Drum
Boiling Point 78.37°C
Percent Purity Min 94.9% (v/v)
Grade FCC
Chemical Name or Material Ethanol
ShelfLife 36 Months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.