Learn More
Abnova™ Gram Bacteria DNA Purification Kit (Pre-filled)
Catalog No. 89339134 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
89-339-134 1 Kit
1 options
Catalog No. 89-339-134 Supplier Abnova™ Supplier No. KA5512
Add to Cart
Add to Cart
Gram Bacteria DNA Purification Kit (Pre-filled) is designed specifically for the optimal automated extraction of DNA from gram bacteria on the SpiralPipet™ instrument.
Specifications
Content And Storage Store at room temperature.
Isolation Technology Magnetic Bead
Product Type Magnetic Bead Handling system
Quantity 1 Kit