Learn More
Newcomer Supply Gram, Brown-Brenn Modified Stain Kit, 250 ml kit
SDP
Catalog No. NC0441715
Encompass
Newcomer Supply Gram, Brown-Brenn Modified Stain Kit is a simple and rapid procedure for differential staining of gram-positive and gram-negative bacteria with a tartrazine counterstain. No Picric Acid!
Catalog No. NC0441715 Supplier Newcomer Supply Supplier No. 9123A
Newcomer Supply Gram, Brown-Brenn Modified Stain Kit is a simple and rapid procedure for differential staining of gram-positive and gram-negative bacteria with a tartrazine counterstain. No Picric Acid!