Learn More
Newcomer Supply Gram, Brown-Hopps Stain Kit, 250 ml kit
SDP
Catalog No. NC0441713
Encompass
Newcomer Supply Gram, Brown-Hopps Stain Kit procedure, is a superior and consistently reliable Gram stain that uses Gallego Solution to differentiate and fix staining of gram-negative and gram-positive bacteria in tissue sections.
Catalog No. NC0441713 Supplier Newcomer Supply Supplier No. 9124A
Newcomer Supply Gram, Brown-Hopps Stain Kit procedure, is a superior and consistently reliable Gram stain that uses Gallego Solution to differentiate and fix staining of gram-negative and gram-positive bacteria in tissue sections.