Learn More
StatLab Gram Control Slides
Description
- Locate smeared area easily within clearly defined wells that will not cross-contaminate patient wells
- Control Slides are stained with Gram Crystal violet, iodine, acetone-alcohol decolorizer, and then counterstained with Safranin
- 50 slides/box
Specifications
Specifications
| Product Type | Control Slide |
| Quantity | 50 Slides/Box |