Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Gram Crystal Violet
GSA_VA

Catalog No. B12525 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
3.8 L
4 x 250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12525 4 x 250mL
B12526 3.8 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B12525 Supplier BD Supplier No. 212525
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Can be used as a replacement reagent for the BD™ Gram Stain Kits. BD™ Gram Crystal Violet is used as the primary stain in a 4-step Gram staining process. Available in 250ml solution bottles for direct use or gallon containers for bulk storage.

Gram Stain reagents are used to stain microorganisms from cultures or specimens by the differential Gram method.

  • Complete stain kits reduce the mess associated with Gram staining procedures and include a compact tray for holding the 250mL reagent bottles
  • Break-resistant bottles features no-drip, keep-clean cap spouts that make regulating the flow easy and that help to prevent spots and spills
  • Color-coded labels allow quick identification of the reagent

Order Info

Available in packs of four 250mL bottles or individual one-gallon bottles

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 4 x 250mL
Certifications/Compliance CE
Format Liquid
For Use With (Application) Used to stain microorganisms from cultures or specimens by the differential gram method
Color Purple
Storage Requirements 15° to 30°C
Stain Type Crystal Violet
Volume (Metric) 250 mL
Product Type Crystal Violet
Packaging Direct use reagent bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.