Learn More
StatLab Gram Positive/Gram Negative Control Slides
SDP
Catalog No. 21033335
Change view
Click to view available options
Includes:
Gram
Gram Pos/Neg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
21-033-335 Gram Pos/Neg
21-033-336 Gram
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-033-335 Supplier StatLab Supplier No. CSG0225P
Only null left
Add to Cart
Add to Cart
Use gram positive/gram negative control slides with confidence to evaluate stain quality and performance in histology laboratories
  • Slides consist of formalin fixed paraffin embedded tissue sections which contain known distinctive tissue elements
  • Use these slides with confidence to evaluate stain quality and performance in histology laboratories
  • Includes 25 Unstained Slides per pack
Specifications
Includes Gram Pos/Neg
Quantity 25 Pk.
Type Gram Stain Slide