Learn More
Volu Sol Gram's Iodine Solution Stabilized, 128 oz / 3.78 L, Volu-Sol™
SDP
Supplier:  Volu Sol VGI128
Encompass_Preferred
Iodine Solution Stabilized, component of the Gram Stain Kit.  Gram's Iodine acts as a mordant in the Gram procedure, fixing the crystal violet to cell walls
Catalog No. 50-420-804