Learn More
Volu Sol Gram's Iodine Solution Stabilized, 5 gal / 20 L, Volu-Sol™
Supplier: Volu Sol VGI640
Iodine Solution Stabilized, component of the Gram Stain Kit. Gram's Iodine acts as a mordant in the Gram procedure, fixing the crystal violet to cell walls