Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gram's Iodine, dilute Lugol's Iodine, Ricca Chemical
SDP

Catalog No. p-3251732 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging Type:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
R3450000100 100 mL Amber Glass Bottle
3450-16 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R3450000100 Supplier Ricca Chemical Company Supplier No. R3450000100C
Only null left
Add to Cart
Add to Cart

For Gram Staining

  • For Gram Staining

Specifications

Type Gram Iodine, Lugol's Iodine
Quantity 100 mL
Color Brown
Packaging Type Amber Glass Bottle
Stain Type Dilute Lugol's Iodine
Volume (Metric) 100 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.