Learn More
Gram's Iodine Solution, Sigma-Aldrich
Catalog No. 01258833 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2.5 L
Catalog No. Quantity
01-258-833 2.5 L
1 options
Catalog No. 01-258-833 Supplier MilliporeSigma Supelco Supplier No. 901072.5LF
Add to Cart
Add to Cart
Mordant solution used to form crystal violet complexes for Gram staining in microbiology applications.
Gram's iodine solution is used as a mordant in the Gram staining process. It is added after the primary staining step with Crystal Violet; the resultant Crystal Violet - Iodine complex imparts purple color to the cells.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 2.5 L
Product Type Gram Iodine